Stampa…
Anti-MUC5AC Mouse Monoclonal Antibody (CF640R) [clone: CLH2]
Fornitore: Biotium
BNC400897-500 BNC400897-100
BNC400897-500EA 560 EUR
BNC400897-500 BNC400897-100
Anti-MUC5AC Mouse Monoclonal Antibody (CF640R) [clone: CLH2]
Anticorpi
Mucin 5AC glycoprotein (MUC5AC) is a HMW glycoprotein belonging to the superfamily of mucins. Mucins are produced by epithelial cells and can be divided into two families; secretory mucins and membrane bound mucins. MUC5AC is a mucus-forming, secreted mucin that is found in normal gastric and tracheo-bronchial mucosa, but absent from normal colon. MUC5AC expression is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differentialMore Product Information