Stampa…

Leptin

Fornitore: Biorbyt

ORB97815-100
BIRBORB97815-100EA 0 EUR
BIRBORB97815-100
Leptin
Proteine e Peptidi

Visualizza i codici

Specifiche

Protein/peptide name Leptin
Environmentally Preferable
Protein synonyms LEP
Specie
Sorgente luminosa
Attività biologica This Leptin triple mutant is capable of inhibiting leptininduced proliferation of BAF/3 cells stably transfectedwith the long form of human leptin receptor. It also inhibitsvarious leptin effects in several in vitro bioassays.
Purezza 95% pure by SDS-PAGE
Formulazione Lyophilized. Leptin is lyophilized from a solution containing 0.003 mM NaHCO3.
Peso molecolare 16 kDa
Condizioni di conservazione Store at−20 °C.
Solubilità It is recommended to reconstitute the lyophilized Leptinmutant in sterile 0.4% NaHCO3 adjusted to pH 8-9, notless than 100 g/ml, which can then be further diluted toother aqueous solutions. At low concentrations additionof a carrier protein (0.1% HSA or B

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR