Stampa…
Real Time PCR Detetction Test Kits, SUPREME
Fornitore: BIOPREMIER
SFS-0059 SFS-0006 SFS-0007 SFS-0008 SFS-0009 SFS-0010 SFS-0011 SFS-0012 SFS-0013 SFS-SP0006 SFS-SP0007 SFS-SP0008 SFS-SP0009 SFS-SP0010 SFS-SP0011 SFS-SP0012 SFS-SP0013 SFS-056 SFS-057 SFS-058 SFS-SP056 SFS-SP057 SFS-SP058 SFS-SP0059 SFS-0066 SFS-SP0066 SFS-0067 SFS-SP0067
BIOPSFS-0059EA 673 EUR
BIOPSFS-0059 BIOPSFS-0006 BIOPSFS-0007 BIOPSFS-0008 BIOPSFS-0009 BIOPSFS-0010 BIOPSFS-0011 BIOPSFS-0012 BIOPSFS-0013 BIOPSFS-SP0006 BIOPSFS-SP0007 BIOPSFS-SP0008 BIOPSFS-SP0009 BIOPSFS-SP0010 BIOPSFS-SP0011 BIOPSFS-SP0012 BIOPSFS-SP0013 BIOPSFS-056 BIOPSFS-057 BIOPSFS-058 BIOPSFS-SP056 BIOPSFS-SP057 BIOPSFS-SP058 BIOPSFS-SP0059 BIOPSFS-0066 BIOPSFS-SP0066 BIOPSFS-0067 BIOPSFS-SP0067
Real Time PCR Detetction Test Kits, SUPREME
Test per microbiologia Test PCR
SUPREME Real time detection uses real-time PCR for the detection of DNA present in the mitochondrial genome in a simple, reliable, and rapid procedure. The assay is based on 5’ nuclease real time PCR reactions to amplify a unique genomic sequence in the target.
  • Amplification and detection of DNA
  • Real-time PCR with rapid hot-start Taq DNA polymerase
  • Internal Control to exclude false-negative results
  • Optimised to handle PCR inhibitors, containing UDG enzyme
  • Higher stability, enzymes in seperate tubes
  • PCR- platforms: runs on all established standard real-time PCR- platforms
  • Harmonised thermal profiles to run with other SUPREME BIOPREMIER kits simultaneously
  • Channel Target: FAM, Channel IC: ROX
  • Available formats: Kits with 100 or 10 reactions
Our carefully designed primers and probe ensure highest sensitivity and specificity. The kit consists of an assay mix for the detection of the target as well as a positive control and an internal control (IC). To minimise PCR cross-contamination, the Kit contains dUTP and uracil-N glycosylase (UDG). Comparative quantification analysis of DNA in food and feedstuff products to estimate the percentage of the fraudulent products.