Stampa…
AutoCollect™ Specimen Collection Cards
Fornitore: Ahlstrom-Munksjö
8.560.0003.B-N 8.295.0003.B-N 8.566.0004.B-N 8.560.0006.B-N 8.567.0006.B-N 8.296.0003.B-N 8.561.0004.B-N
BINZ8.560.0003.B-NEA 451 EUR
BINZ8.560.0003.B-N BINZ8.295.0003.B-N BINZ8.566.0004.B-N BINZ8.560.0006.B-N BINZ8.567.0006.B-N BINZ8.296.0003.B-N BINZ8.561.0004.B-N
AutoCollect™ Specimen Collection Cards
Sistemi di diagnostica Raccolta/Preparazione di Campioni Clinici Biological Sample Collection/Preparation
Ahlstrom AutoCollect™ specimen collection cards for high-throughput environments work efficiently in both DBS punch and direct elution. They are designed for microvolume sampling, transport and storage of dried blood spot (DBS) at ambient temperature.
  • Pharmacokinetic
  • Therapeutic drugs monitoring
  • Preclinical and clinical studies
  • Human identification
  • Toxicology
  • Forensics
  • Biomarkers screening
  • Viral load testing
The cassette format of the card with high stiffness provides a suitable fit with main laboratory automated equipment available on the market (More Product Information