Stampa…
Cleanroom Basic Chairs
Fornitore: bimos
9145-2571 9142-2571 9146-2571 9143-2571 9140-2571-812 9140-2571-807 91416902525807
139-0582EA 618 EUR
139-0582 139-0580 139-0583 139-0581 BIMO9140-2571-812 BIMO9140-2571-807 BIMO91416902525807
Cleanroom Basic Chairs
Sedie
In order to create 'clean' or 'sterile' rooms, measures have to be implemented to prevent products and processes from becoming contaminated. This is a key requirement of the microelectronics sector, the pharmaceuticals industry, microsystem production, optics, medical engineering and healthcare. That is why bimos has worked together with industry and research specialists to develop its innovative range of cleanroom chairs for use in extreme conditions.
  • The smooth, sealed surfaces of the seat and backrest shells prevent swirl even when there is a constant