Stampa…
Safetyspace® filter tips, 10 - 1000 µl, Extended ST (10x96)
Fornitore: Sartorius
LH-XF791001
BHITLH-XF791001EA 188 EUR
BHITLH-XF791001
Safetyspace® filter tips, 10 - 1000 µl, Extended ST (10x96)
Puntali per pipette
The Safetyspace Extended Plus filtered tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware. The tip design allows for more efficient retrieval of samples, down to the last drop. The longer length of these tips helps prevent contamination by ensuring that the pipette does not come into contact with the walls of labware or containers during use. The filter provides an additional layer of protection against sample carryover, which also contributes to reducing the need for frequent pipette maintenance.
  • Maximized reach
  • Added protection