Stampa…
Warming plate, 37 °C, Bel-Art ProCulture, SP Bel-Art
Fornitore: Bel-Art Products, a Part of SP
F370151000
BELAF370151000EA 1370 EUR
BELAF370151000
Warming plate, 37 °C, Bel-Art ProCulture, SP Bel-Art
Termostati Piastre riscaldanti
Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37 °C (±1 °C) outside of the incubator.
  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Can be used to maintain the warmth of reagents inside the hood
  • Fits easily in cell culture hoods or on the workbench
  • Temperature remains uniform across the entire anodized aluminum warming plate
  • 15-minute count down timer keeps track of time sensitive applications
  • UV resistant polypropylene housing will not break down under