Stampa…
Cryo Storage Box −20 °C Cryo-Safe™ Maxi Cooler, SP Bel-Art
Fornitore: Bel-Art Products, a Part of SP
F188462020
BELAF188462020EA 545 EUR
BELAF188462020
Cryo Storage Box −20 °C Cryo-Safe™ Maxi Cooler, SP Bel-Art
Scatole Sistema di raffreddamento da banco
Polycarbonate cooler body and lid, gel-filled.
  • Adapters change well size to hold 0,5 ml microcentrifuge tubes
  • Small footprint is ideal for benchtop use
  • Epoxy coated wire handle fastens lid and allows easy transport
  • All polycarbonate cooler body and lid clean easily
A non-toxic freezing gel is isolated within the walls of the cooler and lid, ensuring lasting low temperatures and protecting any labels or marks on the vials. The lid is indexed and marked to show tube location that matches locations printed on the cooler. Coolers can be stacked to saveMore Product Information