Stampa…
Funnels, powder, with tapered stem, SP Bel-Art
Fornitore: Bel-Art Products, a Part of SP
F146810000
BELAF146810000EA 1330 EUR
BELAF146810000
Funnels, powder, with tapered stem, SP Bel-Art
Imbuti Imbuti per polveri
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable at 121 °C
With standard tapered stem.