Stampa…
Wash bottles, wide neck, colour coded, safety labelled, SP Bel-Art
Fornitore: Bel-Art Products, a Part of SP
F117160009 F117160002 F117160001 F117160011 F117160015 F117160020 F117160004 F117320009 F117320019
BELAF117160009EA 85.5 EUR
BELAF117160009 BELAF117160002 BELAF117160001 BELAF117160011 BELAF117160015 BELAF117160020 BELAF117160004 BELAF117320009 BELAF117320019
Wash bottles, wide neck, colour coded, safety labelled, SP Bel-Art
Bottiglie Spruzzette
These wash bottles, made of LDPE, translucent, with PP spray cup feature a 53 mm wide mouth for easy refilling and are pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) section 704 four-colour diamond, health hazards, Chemical Abstract System (CAS) number and suggested protective clothing and equipment. Leak-proof, these bottles have PP polypropylene closures to aid in quick and easy identification.
  • Good chemical
  • Wide neck for quick, safe filling
  • Colour-coded closures for easy identification