Stampa…
CytoFLEX Mosaic Detection Modules
Fornitore: Beckman Coulter
D19589 D20166 D20162 D20161 D19581 D20160 D19586 D20164 D19585 D20165 D20163 D19583 D19584 D19582
BECMD19589EA 5410 EUR
BECMD19589 BECMD20166 BECMD20162 BECMD20161 BECMD19581 BECMD20160 BECMD19586 BECMD20164 BECMD19585 BECMD20165 BECMD20163 BECMD19583 BECMD19584 BECMD19582
CytoFLEX Mosaic Detection Modules
Conta cellule Citometri a flusso
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
  • A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful