Stampa…

Barnstead™ E-Pure™ Ultrapure Water Systems

Fornitore: Thermo Fisher Scientific
D4632-33 D4642-33 D4641
BARND4632-33EA 4020 EUR
BARND4632-33 BARND4642-33 BARND4641
Barnstead™ E-Pure™ Ultrapure Water Systems
Purificatori d'acqua Sistemi per acqua pura e ultrapura
The Barnstead™ E-Pure™ is designed to connect directly to the tap water feed, and it can handle all of the laboratory high volume ultrapure water applications with low operating costs. This system is ideal for AA, GS, buffer preps, or as a purified feed water source for analytical instruments or labware washers.

  • Produce Type 1 ultrapure water with low operating costs
  • It is simple to use and easy to maintain
  • Virgin polypropylene water pathways prevent recontamination
  • System is wall-mountable and completely assembled for easy installation
  • Quarter-turn quick
More Product Information
Visualizza i codici

Specifiche

Modello D4632-33 D4642-33 D4641
Environmentally Preferable
Acqua d’alimentazione Tap or pretreated, 6,9 bar Tap or pretreated, 6,9 bar Tap or pretreated, 6,9 bar
Portata Up to 2,5 L/min Up to 2,5 L/min Up to 2,0 L/min
Conducibilità 0,055 μS/cm 0,055 μS/cm 0,055 μS/cm
Contenuto TOC <10 ppb <10 ppb <10 ppb
Resistività a +25 °C 18,2 MΏ.cm 18,2 MΏ.cm 18,2 MΏ.cm
Batteri <1 CFU/ml <1 CFU/ml <1 CFU/ml
Particelle <1 μm/ml <1 μm/ml <1 μm/ml
Elettrico 240 V 240 V 120 V
L×P×A 740×190×710 mm 953×190×711 mm 889×190×711 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR