Stampa…
Cryogenic dual storage vesssel, Arctic Express®
Fornitore: Thermo Fisher Scientific
CK50920 CK50921 CK50922
BARNCK50920EA 2390 EUR
BARNCK50920 BARNCK50921 BARNCK50922
Cryogenic dual storage vesssel, Arctic Express®
Dewar e vasi criogenici
Dual storage vessels are used to protect non-hazardous samples during shipment and storage. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
  • Aluminum vessels with lockable lid hold six numbered, colour-coded, stainless-steel cans, which accommodate canes that hold ampules
When filled with liquid nitrogen and placed in the laboratory, unit can be used as a 'can and cane' storage system indefinitely with static holding times of up to 125 days.
Informazioni per l'ordine: More Product Information