Stampa…
Water purification systems, Barnstead™ LabTower™ EDI
Fornitore: Thermo Fisher Scientific
50132395 50132396
BARN50132395EA 18900 EUR
BARN50132395 BARN50132396
Water purification systems, Barnstead™ LabTower™ EDI
Purificatori d'acqua Sistemi per acqua pura e ultrapura
This system combines purification with EDI technology and storage in one. Users can conveniently dispense Type 1 water directly from the system while Type 2 water is accessible from the integrated 100 l reservoir, which is an ideal supply for lab equipment such as autoclaves.
  • Unique system with pre-treatment plus polisher to produce both Type 1 and Type 2 water
  • Compact, free-standing unit can be easily relocated with bottom-mounted rollers
  • Validated 0,2 µm final filter with folded membrane can be autoclaved up to five times
  • Standard UV photo-oxidation at 18