Stampa…

Aminoacetonitrile sulphate 97%

Fornitore: Thermo Fisher Scientific
Pericolo

B22155.14 B22155.22 B22155.36
B22155.14EA 30.4 EUR
B22155.14 B22155.22 B22155.36
Aminoacetonitrile sulphate 97%
Aminoacetonitrile sulphate

Purity: 97% CAS No.: 5466-22-8 Molecular Formula: C4H8N8.H2SO4 Formula Weight: 210.21 Odour: Odourless Melting Point: ca 160° dec. Beilstein Reference: 3916094 UN No.: UN3439 MDL No.: MFCD00012851

Formula: C₄H₁₀N₄O₄S
Punto di fusione: ∼ 160 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012851
CAS Number: 5466-22-8
EINECS: 226-772-0
ADR: 6.1,III
Merck Index: 14,412

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR