Stampa…
Multi-channel pipettes, variable volume, Axygen® Axypet™
Fornitore: Corning
AP-12-10 AP-12-200 AP-12-300 AP-12-50 AP-8-10 AP-8-200 AP-8-300 AP-8-50
AXYGAP-12-10EA 732 EUR
AXYGAP-12-10 AXYGAP-12-200 AXYGAP-12-300 AXYGAP-12-50 AXYGAP-8-10 AXYGAP-8-200 AXYGAP-8-300 AXYGAP-8-50
Multi-channel pipettes, variable volume, Axygen® Axypet™
Pipette Pipette automatiche
These pipettes combine the highest levels of accuracy and precision with design innovations intended to make the pipetting experience more comfortable over prolonged periods.
  • Digital volume display with easy to read numbers
  • Advanced, lightweight materials (body of the pipette and piston assemblies)
  • Each channel has an individual precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next, as well as between channels
  • Micrometer is continuously adjustable for selection of whole or fractional volumes
The plungerMore Product Information