Stampa…

Sodio cloruro Food

Fornitore: PanReac AppliChem

Synonyms: Saline, NaCl, Sea salt, Halite

206994.0416 206994.1214
APPC206994.0416EA 0 EUR
APPC206994.0416 APPC206994.1214
Sodio cloruro Food
Sodio cloruro
Formula: NaCl
Peso molecolare: 58.44 g/mol
Punto di ebollizione: 1413 °C
Punto di fusione: 804 °C
Densità: 2,16 g/cm³ at 20 °C
Temperatura di stoccaggio: Room Temperature
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Visualizza i codici

Risultati del test di specificazione

Assay (as NaCl) after drying at 625 °C 99.0-100.5%
Identity: Chloride passes test
Sodium passes test
Appearance passes test
Loss on drying not more than: 0.5%
Ferrocyanide (as Na4 Fe(CN)6) not more than: 0.0013 %
Calcium and Magnesium not more than: 0.35%
Barium not more than: 300 ppm
Sodium nitrite not more than: 0.5 %
Arsenic (as As) not more than: 1 ppm
Heavy metals (as Pb) not more than: 2 ppm
For use in foodstuffs according to F.C.C. Specifications F.C.C. 12


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR