Stampa…

Sodio cloruro BP, JP, Ph. Eur., USP, puro

Fornitore: PanReac AppliChem

141659.0416 141659.1211 141659.1214
APPC141659.0416EA 167 EUR
APPC141659.0416 APPC141659.1211 APPC141659.1214
Sodio cloruro BP, JP, Ph. Eur., USP, puro
Sodio cloruro
Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR