Stampa…

Metanolo ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO per analisi

Fornitore: PanReac AppliChem
Pericolo

131091.1212 131091.0716 131091.1211 131091.1214 131091.1611 131091.1612
APPC131091.1212EA 90.1 EUR
APPC131091.1212 APPC131091.0716 APPC131091.1211 APPC131091.1214 APPC131091.1611 APPC131091.1612
Metanolo ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO per analisi
Metanolo

For analysis, ACS, and ISO.

Formula: CH₃OH
Punto di ebollizione: 64,..65 °C
Punto di fusione: –97,8 °C
Densità: 0,792 kg/l
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Minimum Assay (G.C.) 99.9%
Identity IR Passes Test
Density 20/4 0.791 - 0.79
APHA colour 10
Acidity 0.0002 meq/g
Alkalinity 0.0002 meq/g
Insoluble Matter in H₂O Passes Test
Non-Volatile Matter 0.0005%
Reducing Substance to KMnO₄ (as O) 0.00025%
Darkened Substances by H₂SO₄ Passes Test
Carbonyl Compounds (as CH₃COCH₃) 0.002%
Acetaldehyde (CH₃CHO) 0.001%
Acetone (G.C.) 0.001%
Formaldehyde (HCHO) 0.001%
Ethanol (G.C.) 0.005%
2-Propanol (G.C.) 0.005%
Water(H₂O) 0.05%
Metals by ICP, Ag 0.05 mg/kg (ppm)
Metals by ICP, Al 0.5 mg/kg (ppm)
Metals by ICP, As 0.05 mg/kg (ppm)
Metals by ICP, Au 0.05 mg/kg (ppm)
Metals by ICP, B 0.02 mg/kg (ppm)
Metals by ICP, Ba 0.1 mg/kg (ppm)
Metals by ICP, Be 0.02 mg/kg (ppm)
Metals by ICP, Bi 0.05 mg/kg (ppm)
Metals by ICP, Ca 0.1 mg/kg (ppm)
Metals by ICP, Cd 0.05 mg/kg (ppm)
Metals by ICP, Co 0.02 mg/kg (ppm)
Metals by ICP, Cr 0.02 mg/kg (ppm)
Metals by ICP, Cu 0.02 mg/kg (ppm)
Metals by ICP, Fe 0.1 mg/kg (ppm)
Metals by ICP, Ga 0.02 mg/kg (ppm)
Metals by ICP, Ge 0.05 mg/kg (ppm)
Metals by ICP, Hg 0.05 mg/kg (ppm)
Metals by ICP, In 0.05 mg/kg (ppm)
Metals by ICP, K 0.1 mg/kg (ppm)
Metals by ICP, Li 0.05 mg/kg (ppm)
Metals by ICP, Mg 0.05 mg/kg (ppm)
Metals by ICP, Mn 0.02 mg/kg (ppm)
Metals by ICP, Mo 0.02 mg/kg (ppm)
Metals by ICP, Na 0.5 mg/kg (ppm)
Metals by ICP, Ni 0.02 mg/kg (ppm)
Metals by ICP, P 0.2 mg/kg (ppm)
Metals by ICP, Pb 0.02 mg/kg (ppm)
Metals by ICP, Pt 0.02 mg/kg (ppm)
Metals by ICP, S 0.2 mg/kg (ppm)
Metals by ICP, Sb 0.02 mg/kg (ppm)
Metals by ICP, Si 0.2 mg/kg (ppm)
Metals by ICP, Sn 0.1 mg/kg (ppm)
Metals by ICP, Sr 0.2 mg/kg (ppm)
Metals by ICP, Ti 0.02 mg/kg (ppm)
Metals by ICP, Tl 0.02 mg/kg (ppm)
Metals by ICP, V 0.02 mg/kg (ppm)
Metals by ICP, Zn 0.1 mg/kg (ppm)
Metals by ICP, Zr 0.02 mg/kg (ppm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR