Stampa…
Terrestrial (Cypripedium) orchid medium, contains 600 mg/L calcium nitrate
Fornitore: Apollo Scientific
PMT842A-10X1L PMT842B-10L PMT842C-50L
APOSPMT842A-10X1LEA 32.2 EUR
APOSPMT842A-10X1L APOSPMT842B-10L APOSPMT842C-50L
Terrestrial (Cypripedium) orchid medium, contains 600 mg/L calcium nitrate
Terreni per colture cellulari
Contains 600 mg/L calcium nitrate and 200 mg/L casein; without ammonium nitrate.
  • Plant tissue culture tested