Stampa…

2-Fluoro-3-(hydroxymethyl)phenol 95%

Fornitore: Apollo Scientific

PC910231-1G
APOSPC910231-1GEA 632 EUR
APOSPC910231-1G
2-Fluoro-3-(hydroxymethyl)phenol 95%
2-Fluoro-3-(hydroxymethyl)phenol
Formula: C₇H₇FO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12923294
CAS Number: 960001-66-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR