Stampa…

Methyl 3-fluoro-4-(methylsulphonyl)benzoate

Fornitore: Apollo Scientific
Attenzione

PC56035-1G PC56035-5G PC56035-25G
APOSPC56035-1GEA 111 EUR
APOSPC56035-1G APOSPC56035-5G APOSPC56035-25G
Methyl 3-fluoro-4-(methylsulphonyl)benzoate
Methyl 3-fluoro-4-(methylsulphonyl)benzoate
Formula: C₉H₉FO₄S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD15143295
CAS Number: 1215074-49-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR