Stampa…

4-(Difluoromethoxy)-3-hydroxyphenylacetic acid

Fornitore: Apollo Scientific

PC510305-1G
APOSPC510305-1GEA 1010 EUR
APOSPC510305-1G
4-(Difluoromethoxy)-3-hydroxyphenylacetic acid
4-(Difluoromethoxy)-3-hydroxyphenylacetic acid
Formula: C₉H₈F₂O₄
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD29065905
CAS Number: 1312884-10-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR