Stampa…

4-Trifluoromethyl-L-phenylalanine 95%

Fornitore: Apollo Scientific

PC4463-10G PC4463-1G PC4463-5G
APOSPC4463-10GEA 800 EUR
APOSPC4463-10G APOSPC4463-1G APOSPC4463-5G
4-Trifluoromethyl-L-phenylalanine 95%
4-Trifluoromethyl-L-phenylalanine
Formula: C₁₀H₁₀F₃NO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00044946
CAS Number: 114926-38-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR