Stampa…

2,6-Dichloro-4-(trifluoromethoxy)phenylacetic acid 97%

Fornitore: Apollo Scientific

PC3698-1G PC3698-5G
APOSPC3698-1GEA 161 EUR
APOSPC3698-1G APOSPC3698-5G
2,6-Dichloro-4-(trifluoromethoxy)phenylacetic acid 97%
2,6-Dichloro-4-(trifluoromethoxy)phenylacetic acid
Formula: C₉H₅Cl₂F₃O₃
Temperatura di stoccaggio: Ambiente
CAS Number: 886503-16-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR