Stampa…

2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetic acid

Fornitore: Apollo Scientific

PC303555-1G PC303555-5G
APOSPC303555-1GEA 430 EUR
APOSPC303555-1G APOSPC303555-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetic acid
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetic acid
Formula: C₉H₅BrF₄O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD32640133
CAS Number: 2385550-69-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR