Stampa…

2-Amino-3'-fluorobiphenyl hydrochloride

Fornitore: Apollo Scientific
Attenzione

PC11124-1G
APOSPC11124-1GEA 270 EUR
APOSPC11124-1G
2-Amino-3'-fluorobiphenyl hydrochloride
2-Amino-3'-fluorobiphenyl hydrochloride
Formula: C₁₂H₁₀FN·ClH
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06795781
CAS Number: 139769-18-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR