Stampa…

Diallylamine hydrochloride 95%

Fornitore: Apollo Scientific

OR939744-25G
APOSOR939744-25GEA 84.1 EUR
APOSOR939744-25G
Diallylamine hydrochloride 95%
Diallylamine hydrochloride
Formula: C₆H₁₁N
Peso molecolare: 133.62 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00060150
CAS Number: 6147-66-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR