Stampa…

2-Isobutyl-3-methylpyrazine 95%

Fornitore: Apollo Scientific

OR938702-25G
APOSOR938702-25GEA 71.2 EUR
APOSOR938702-25G
2-Isobutyl-3-methylpyrazine 95%
2-Isobutyl-3-methylpyrazine
Formula: C₉H₁₄N₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00059770
CAS Number: 13925-06-9

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR