Stampa…

N-Decanoylsarcosine sodium salt 98%

Fornitore: Apollo Scientific

OR936414-1G OR936414-5G
APOSOR936414-1GEA 174 EUR
APOSOR936414-1G APOSOR936414-5G
N-Decanoylsarcosine sodium salt 98%
N-Decanoylsarcosine sodium salt
Formula: C₁₃H₂₅NO₃
Punto di fusione: 155 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00451533
CAS Number: 30377-07-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR