Stampa…

4-tert-Butoxybenzaldehyde 95%

Fornitore: Apollo Scientific

OR936350-1G OR936350-25G OR936350-5G
APOSOR936350-1GEA 31.9 EUR
APOSOR936350-1G APOSOR936350-25G APOSOR936350-5G
4-tert-Butoxybenzaldehyde 95%
4-tert-Butoxybenzaldehyde
Formula: C₁₁H₁₄O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00040800
CAS Number: 57699-45-3

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR