Stampa…

4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) 95%

Fornitore: Apollo Scientific

OR930649-25G OR930649-5G
APOSOR930649-25GEA 464 EUR
APOSOR930649-25G APOSOR930649-5G
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) 95%
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Formula: C₁₁H₂₀O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00042622
CAS Number: 5451-55-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR