Stampa…

Isopropyl cyclohexanecarboxylate 95%

Fornitore: Apollo Scientific

OR921446-100G OR921446-25G
APOSOR921446-100GEA 377 EUR
APOSOR921446-100G APOSOR921446-25G
Isopropyl cyclohexanecarboxylate 95%
Isopropyl cyclohexanecarboxylate
Formula: C₁₀H₁₈O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00058949
CAS Number: 6553-80-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR