Stampa…

1,4-Benzoquinone dioxime 95%

Fornitore: Apollo Scientific

OR920023-100G OR920023-250G OR920023-25G
APOSOR920023-100GEA 226 EUR
APOSOR920023-100G APOSOR920023-250G APOSOR920023-25G
1,4-Benzoquinone dioxime 95%
1,4-Benzoquinone dioxime
Formula: C₆H₆N₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00063636
CAS Number: 105-11-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR