Stampa…

3-Propoxytoluene 98%

Fornitore: Apollo Scientific

OR914596-10G OR914596-1G OR914596-5G
APOSOR914596-10GEA 806 EUR
APOSOR914596-10G APOSOR914596-1G APOSOR914596-5G
3-Propoxytoluene 98%
3-Propoxytoluene
Formula: C₁₀H₁₄O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05982065
CAS Number: 33426-65-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR