Stampa…

(±)-3-Chloromandelic acid 98%

Fornitore: Apollo Scientific

OR911512-100G OR911512-25G
APOSOR911512-100GEA 237 EUR
APOSOR911512-100G APOSOR911512-25G
(±)-3-Chloromandelic acid 98%
(±)-3-Chloromandelic acid
Formula: C₈H₇ClO₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00829409
CAS Number: 16273-37-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR