Stampa…

2-Isopropoxy-2-phenylacetic acid 97%

Fornitore: Apollo Scientific

OR907836-1G OR907836-250MG
APOSOR907836-1GEA 400 EUR
APOSOR907836-1G APOSOR907836-250MG
2-Isopropoxy-2-phenylacetic acid 97%
2-Isopropoxy-2-phenylacetic acid
Formula: C₁₁H₁₄O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16661867
CAS Number: 5394-87-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR