Stampa…

D-(+)-Glucosamine hydrochloride 98+%

Fornitore: Apollo Scientific
Attenzione

Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

OR5282-1KG OR5282-250G OR5282-5KG
APOSOR5282-1KGEA 52.2 EUR
APOSOR5282-1KG APOSOR5282-250G APOSOR5282-5KG
D-(+)-Glucosamine hydrochloride 98+%
D-(+)-Glucosamine hydrochloride
Formula: C₆H₁₃NO₅
Peso molecolare: 215.63 g/mol
Punto di fusione: ∼ 192 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00135831
CAS Number: 66-84-2
EINECS: 200-638-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR