Stampa…

2-Cyanobenzenecarboximidamide hydrochloride

Fornitore: Apollo Scientific

Synonyms: 2-Cyanobenzamidine hydrochloride

OR470985-1G
APOSOR470985-1GEA 397 EUR
APOSOR470985-1G
2-Cyanobenzenecarboximidamide hydrochloride
2-Cyanobenzenecarboximidamide hydrochloride
Formula: C₈H₈ClN₃
Peso molecolare: 181.63 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16622254
CAS Number: 426828-36-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR