Stampa…

Methyl 2-Amino-4-(1-imidazolyl)benzoate

Fornitore: Apollo Scientific

OR470171-1G OR470171-5G
APOSOR470171-1GEA 199 EUR
APOSOR470171-1G APOSOR470171-5G
Methyl 2-Amino-4-(1-imidazolyl)benzoate
Methyl 2-Amino-4-(1-imidazolyl)benzoate
Formula: C₁₁H₁₁N₃O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18651685
CAS Number: 1272756-25-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR