Stampa…

4-(Benzyloxy)benzyl bromide

Fornitore: Apollo Scientific

OR400483-1G OR400483-250MG
APOSOR400483-1GEA 342 EUR
APOSOR400483-1G APOSOR400483-250MG
4-(Benzyloxy)benzyl bromide
4-(Benzyloxy)benzyl bromide
Formula: C₁₄H₁₃BrO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07368336
CAS Number: 5544-60-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR