Stampa…

(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid 97%

Fornitore: Apollo Scientific

Synonyms: (1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, Boc-D-AcPAC, (1S,3R)-3-(Boc-amino)cyclopentanecarboxylic Acid, (1S,3R)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

OR14678-5G
APOSOR14678-5GEA 253 EUR
APOSOR14678-5G
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid 97%
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid
Formula: C₁₁H₁₉NO₄
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01320857
CAS Number: 261165-05-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR