Stampa…

Isoquinoline-3-carboxylic acid monohydrate 97%

Fornitore: Apollo Scientific
Attenzione

Synonyms: 2-Azanaphthalene-3-carboxylic acid monohydrate, 3-Carboxyisoquinoline monohydrate, Isoquinoline-3-carboxylic acid hydrate

OR0571-1G
APOSOR0571-1GEA 21.9 EUR
APOSOR0571-1G
Isoquinoline-3-carboxylic acid monohydrate 97%
Isoquinoline-3-carboxylic acid monohydrate
Formula: C₁₀H₉NO₃
Peso molecolare: 191.183 g/mol
Punto di fusione: 169…170 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00167216
CAS Number: 203626-75-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR