Stampa…

Methyl [5-(chloroacetyl)-1-methyl-1H-pyrrol-2-yl]acetate

Fornitore: Apollo Scientific

OR01998-1G
APOSOR01998-1GEA 277 EUR
APOSOR01998-1G
Methyl [5-(chloroacetyl)-1-methyl-1H-pyrrol-2-yl]acetate
Methyl [5-(chloroacetyl)-1-methyl-1H-pyrrol-2-yl]acetate
Formula: C₁₀H₁₂ClNO₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09842625
CAS Number: 617721-31-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR