Stampa…

Nichel (II) cloruro esaidrato 99,95%

Fornitore: Apollo Scientific
Pericolo

Synonyms: Nickel dichloride hexahydrate

IN2683-1KG IN2683-250G
APOSIN2683-1KGEA 342 EUR
APOSIN2683-1KG APOSIN2683-250G
Nichel (II) cloruro esaidrato 99,95%
Nichel (II) cloruro esaidrato
Formula: NiCl₂·6H₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149809
CAS Number: 7791-20-0
UN: 3288
ADR: 6.1,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR