Stampa…

Copper(II) dimethyldithiocarbamate 98%

Fornitore: Apollo Scientific
Attenzione

IN1549-25G
APOSIN1549-25GEA 212 EUR
APOSIN1549-25G
Copper(II) dimethyldithiocarbamate 98%
Copper(II) dimethyldithiocarbamate
Formula: C₃H₇NS₂
Punto di fusione: 260 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00050845
CAS Number: 137-29-1
UN: 3077
ADR: 9+EHS,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR