Stampa…

GSK1120212

Fornitore: Apollo Scientific

Synonyms: N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide, Trametinib, JTP-74057

BISN0128-50MG
APOSBISN0128-50MGEA 0 EUR
APOSBISN0128-50MG
GSK1120212
GSK1120212

Trametinib is a highly specific and potent MEK12 inhibitor with IC50 of 0.92 nM1.8 nM, no inhibition of the kinase activities of c-Raf, B-Raf, ERK12.

Formula: C₂₆H₂₃FIN₅O₄
Peso molecolare: 615.4 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD17215075
CAS Number: 871700-17-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR