Stampa…

Streptomicina solfato

Fornitore: Apollo Scientific
Attenzione

BIS0148-100G BIS0148-25G
APOSBIS0148-100GEA 60.9 EUR
APOSBIS0148-100G APOSBIS0148-25G
Streptomicina solfato
Streptomycin sulphate

Streptomycin is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria.


  • Cell culture tested

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Peso molecolare: 1457.4 g/mol
Punto di fusione: 250
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00037023
CAS Number: 3810-74-0
EINECS: 223-286-0
Merck Index: 12,08983

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR