Stampa…

Polymyxin B sulphate

Fornitore: Apollo Scientific
Attenzione

BIP0145-10G BIP0145-1G BIP0145-5G
APOSBIP0145-10GEA 219 EUR
APOSBIP0145-10G APOSBIP0145-1G APOSBIP0145-5G
Polymyxin B sulphate
Polymyxin B sulphate

Poly B is a mixture of the sulphates of polypeptides


  • approx. 8000 units/mg
  • Cell culture tested

Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00131991
CAS Number: 1405-20-5
EINECS: 215-774-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR