Stampa…
HRP-conjugate stabiliser, SeramunStab® Stplus
Fornitore: Apollo Scientific
BIL3111-100ML
APOSBIL3111-100MLEA 62.5 EUR
APOSBIL3111-100ML
HRP-conjugate stabiliser, SeramunStab® Stplus
Sistemi di associazione e marcatura delle proteine
Liquid product colourless to slightly yellow or greenish, odourless, pH 7,3 to 7,5. Stabiliser for conjugates with monomeric HRP.